Assay Details
Gene Symbol: ZFAND4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001302671.1 | NP_001289600.1 | 4 - 5 | 541 | 88 | |
XM_014108758.2 | XP_013964233.1 | 3 - 4 | 279 | 88 | ||
XM_014108759.2 | XP_013964234.1 | 2 - 3 | 206 | 88 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014108759.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
