Assay Details
| Gene Symbol: ECI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022414438.1 | XP_022270146.1 | 4 - 5 | 461 | 81 | |
| XM_535873.6 | XP_535873.3 | 4 - 5 | 600 | 81 | ||
More Information
Set Membership: |
|

ECI2
Genomic Map