Assay Details
| Gene Symbol: PLAAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005639841.3 | XP_005639898.1 | 4 - 5 | 669 | 77 | |
| XM_005639842.3 | XP_005639899.1 | 4 - 5 | 630 | 77 | ||
| XM_005639843.3 | XP_005639900.1 | 4 - 5 | 650 | 77 | ||
| XM_005639844.3 | XP_005639901.1 | 3 - 4 | 505 | 77 | ||
| XM_005639845.3 | XP_005639902.1 | 5 - 6 | 1039 | 77 | ||
| XM_022414182.1 | XP_022269890.1 | 3 - 4 | 449 | 77 | ||
| XM_535846.6 | XP_535846.2 | 4 - 5 | 855 | 77 | ||
More Information
Set Membership: |
|

PLAAT1
Genomic Map