Assay Details
| Gene Symbol: DNAI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005621991.3 | XP_005622048.1 | 3 - 4 | 292 | 81 | |
| XM_014114864.2 | XP_013970339.1 | 3 - 4 | 272 | 81 | ||
| XM_014114865.2 | XP_013970340.1 | 3 - 4 | 275 | 81 | ||
| XM_014114866.2 | XP_013970341.1 | 3 - 4 | 292 | 81 | ||
| XM_022420552.1 | XP_022276260.1 | 3 - 4 | 292 | 81 | ||
| XM_022420553.1 | XP_022276261.1 | 3 - 4 | 292 | 81 | ||
| XM_537092.6 | XP_537092.1 | 3 - 4 | 292 | 81 | ||
More Information
Set Membership: |
|

DNAI3
Genomic Map