Assay Details
| Gene Symbol: PROCA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005624863.3 | XP_005624920.1 | 3 - 4 | 449 | 79 | |
| XM_014116785.2 | XP_013972260.1 | 2 - 3 | 393 | 79 | ||
| XM_014116786.2 | XP_013972261.1 | 2 - 3 | 393 | 79 | ||
| XM_863139.5 | XP_868232.1 | 2 - 3 | 309 | 79 | ||
More Information
Set Membership: |
|

PROCA1
Genomic Map