Assay Details
| Gene Symbol: LYRM9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005624847.3 | XP_005624904.1 | 2 - 3 | 210 | 80 | |
| XM_014116771.2 | XP_013972246.1 | 2 - 3 | 946 | 80 | ||
| XM_014116773.2 | XP_013972248.1 | 2 - 3 | 869 | 80 | ||
| XM_014116774.2 | XP_013972249.1 | 2 - 3 | 1209 | 80 | ||
| XM_022423335.1 | XP_022279043.1 | 3 - 4 | 374 | 80 | ||
| XM_863003.5 | XP_868096.1 | 2 - 3 | 210 | 80 | ||
More Information
Set Membership: |
|

LYRM9
Genomic Map