Assay Details
| Gene Symbol: RNASEH2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022405650.1 | XP_022261358.1 | 2 - 3 | 432 | 69 | |
| XM_022405651.1 | XP_022261359.1 | 2 - 3 | 432 | 69 | ||
| XM_847733.4 | XP_852826.1 | 2 - 3 | 427 | 69 | ||
More Information
Set Membership: |
|

RNASEH2C
Genomic Map