Assay Details
| Gene Symbol: SLC17A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022426253.1 | XP_022281961.1 | 2 - 3 | 348 | 90 | |
| XM_022426254.1 | XP_022281962.1 | 2 - 3 | 396 | 90 | ||
| XM_022426255.1 | XP_022281963.1 | 2 - 3 | 396 | 90 | ||
| XM_532204.6 | XP_532204.2 | 2 - 3 | 395 | 90 | ||
More Information
Set Membership: |
|

SLC17A5
Genomic Map