Assay Details
| Gene Symbol: LOC612942 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022425650.1 | XP_022281358.1 | 5 - 6 | 810 | 77 | |
| XM_022425651.1 | XP_022281359.1 | 6 - 7 | 921 | 77 | ||
| XM_022425652.1 | XP_022281360.1 | 6 - 7 | 926 | 77 | ||
More Information
Set Membership: |
|

LOC612942
Genomic Map