Assay Details
| Gene Symbol: TCP11L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005625952.3 | XP_005626009.1 | 9 - 10 | 1782 | 154 | |
| XM_022424518.1 | XP_022280226.1 | 9 - 10 | 1476 | 154 | ||
More Information
Set Membership: |
|

TCP11L2
Genomic Map