Assay Details
| Gene Symbol: CCDC162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005627693.3 | XP_005627750.1 | 23 - 24 | 4469 | 83 | |
| XM_005627694.3 | XP_005627751.1 | 23 - 24 | 4469 | 83 | ||
| XM_005627695.3 | XP_005627752.1 | 23 - 24 | 4469 | 83 | ||
| XM_022426431.1 | XP_022282139.1 | 23 - 24 | 4471 | 83 | ||
| XM_022426432.1 | XP_022282140.1 | 19 - 20 | 4598 | 83 | ||
More Information
Set Membership: |
|

CCDC162
Genomic Map