Assay Details
| Gene Symbol: CCDC162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005627693.3 | XP_005627750.1 | 27 - 28 | 4984 | 70 | |
| XM_005627694.3 | XP_005627751.1 | 27 - 28 | 4984 | 70 | ||
| XM_005627695.3 | XP_005627752.1 | 27 - 28 | 4984 | 70 | ||
| XM_022426431.1 | XP_022282139.1 | 27 - 28 | 4986 | 70 | ||
| XM_022426432.1 | XP_022282140.1 | 23 - 24 | 5113 | 70 | ||
More Information
Set Membership: |
|

CCDC162
Genomic Map