Assay Details
| Gene Symbol: AKAP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005615511.3 | XP_005615568.1 | 5154 | 70 | ||
| XM_005615512.3 | XP_005615569.1 | 5197 | 70 | |||
| XM_533451.6 | XP_533451.2 | 5306 | 70 | |||
More Information
Set Membership: |
|

AKAP12
Genomic Map