Assay Details
| Gene Symbol: SLC27A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005626491.3 | XP_005626548.1 | 5 - 6 | 1352 | 85 | |
| XM_531894.6 | XP_531894.3 | 6 - 7 | 1542 | 85 | ||
More Information
Set Membership: |
|

SLC27A6
Genomic Map