Assay Details
| Gene Symbol: NDUFB9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005627925.3 | XP_005627982.1 | 353 | 99 | ||
| XM_022426691.1 | XP_022282399.1 | 448 | 99 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: NDUFB9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005627925.3 | XP_005627982.1 | 353 | 99 | ||
| XM_022426691.1 | XP_022282399.1 | 448 | 99 | |||
Set Membership: |
|