Assay Details
| Gene Symbol: DPH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003432004.4 | XP_003432052.1 | 1 - 2 | 346 | 83 | |
| XM_532611.6 | XP_532611.2 | 1 - 2 | 345 | 83 | ||
More Information
Set Membership: |
|

DPH2
Genomic Map