Assay Details
| Gene Symbol: VPS72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005630795.3 | XP_005630852.1 | 2 - 3 | 347 | 59 | |
| XM_533052.6 | XP_533052.1 | 2 - 3 | 348 | 59 | ||
More Information
Set Membership: |
|

VPS72
Genomic Map