Assay Details
| Gene Symbol: SFT2D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014113120.2 | XP_013968595.1 | 1 - 2 | 159 | 78 | |
| XM_533472.6 | XP_533472.2 | 1 - 2 | 159 | 78 | ||
More Information
Set Membership: |
|

SFT2D1
Genomic Map