Assay Details
| Gene Symbol: APOL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005625917.3 | XP_005625974.1 | 5 - 6 | 1706 | 67 | |
| XM_005625918.3 | XP_005625975.1 | 5 - 6 | 1585 | 67 | ||
| XM_014117382.2 | XP_013972857.1 | 5 - 6 | 1698 | 67 | ||
| XM_022424492.1 | XP_022280200.1 | 4 - 5 | 1582 | 67 | ||
| XM_531749.6 | XP_531749.3 | 4 - 5 | 1461 | 67 | ||
More Information
Set Membership: |
|

APOL5
Genomic Map