Assay Details
| Gene Symbol: TXNL4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005615276.3 | XP_005615333.1 | 397 | 87 | ||
| XM_014119994.2 | XP_013975469.1 | 392 | 87 | |||
| XM_022407962.1 | XP_022263670.1 | 403 | 87 | |||
| XM_533363.6 | XP_533363.2 | 371 | 87 | |||
More Information
Set Membership: |
|

TXNL4A
Genomic Map