Assay Details
| Gene Symbol: RPS26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_531628.5 | XP_531628.1 | 261 | 97 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_531628.5 transcripts.
Set Membership: |
|

RPS26
Genomic Map