Assay Details
| Gene Symbol: PACRGL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005618663.3 | XP_005618720.1 | 6 - 7 | 809 | 91 | |
| XM_014112665.2 | XP_013968140.1 | 5 - 6 | 808 | 91 | ||
| XM_014112666.2 | XP_013968141.1 | 6 - 7 | 799 | 91 | ||
| XM_014112667.2 | XP_013968142.1 | 6 - 7 | 694 | 91 | ||
| XM_014112670.2 | XP_013968145.1 | 6 - 7 | 809 | 91 | ||
| XM_022417272.1 | XP_022272980.1 | 5 - 6 | 799 | 91 | ||
| XM_022417273.1 | XP_022272981.1 | 6 - 7 | 685 | 91 | ||
| XM_022417274.1 | XP_022272982.1 | 5 - 6 | 661 | 91 | ||
| XM_022417275.1 | XP_022272983.1 | 6 - 7 | 810 | 91 | ||
| XM_536276.6 | XP_536276.4 | 6 - 7 | 808 | 91 | ||
More Information
Set Membership: |
|

PACRGL
Genomic Map