Assay Details
Gene Symbol: NDUFA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_537861.6 | XP_537861.1 | 2 - 3 | 318 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
