Assay Details
| Gene Symbol: CCT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022421255.1 | XP_022276963.1 | 3 - 4 | 320 | 62 | |
| XM_537245.5 | XP_537245.3 | 5 - 6 | 434 | 62 | ||
| XM_859235.5 | XP_864328.2 | 4 - 5 | 371 | 62 | ||
More Information
Set Membership: |
|

CCT3
Genomic Map