Assay Details
| Gene Symbol: SSR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204756.1 | NP_001191685.1 | 5 - 6 | 478 | 71 | |
| XM_005640995.3 | XP_005641052.1 | 6 - 7 | 839 | 71 | ||
| GenBank mRNA | DN393206.1 | - | 5 - 6 | 478 | 71 | |
More Information
Set Membership: |
|

SSR4
Genomic Map