Assay Details
| Gene Symbol: CCL23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022423297.1 | XP_022279005.1 | 3 - 4 | 361 | 148 | |
| XM_537722.6 | XP_537722.1 | 3 - 4 | 355 | 148 | ||
More Information
Set Membership: |
|

CCL23
Genomic Map