Assay Details
| Gene Symbol: APRT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005620572.3 | XP_005620629.1 | 388 | 63 | ||
| XM_022419087.1 | XP_022274795.1 | 388 | 63 | |||
| XM_536752.5 | XP_536752.2 | 388 | 63 | |||
More Information
Set Membership: |
|

APRT
Genomic Map