Assay Details
| Gene Symbol: CCDC43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005624388.3 | XP_005624445.1 | 2 | 243 | 78 | |
| XM_537617.6 | XP_537617.3 | 1 | 242 | 78 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CCDC43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005624388.3 | XP_005624445.1 | 2 | 243 | 78 | |
| XM_537617.6 | XP_537617.3 | 1 | 242 | 78 | ||
Set Membership: |
|