Assay Details
| Gene Symbol: LOC479795 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_536923.6 | XP_536923.2 | 8 - 9 | 933 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC479795| Gene Symbol: LOC479795 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_536923.6 | XP_536923.2 | 8 - 9 | 933 | 76 | |
Set Membership: |
|