Assay Details
| Gene Symbol: ANXA8L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022417626.1 | XP_022273334.1 | 7 - 8 | 727 | 79 | |
| XM_022417627.1 | XP_022273335.1 | 7 - 8 | 716 | 79 | ||
| XM_536412.6 | XP_536412.2 | 6 - 7 | 624 | 79 | ||
More Information
Set Membership: |
|

ANXA8L1
Genomic Map