Assay Details
| Gene Symbol: MAP7D3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005641869.3 | XP_005641926.1 | 13 - 14 | 2470 | 69 | |
| XM_005641873.3 | XP_005641930.1 | 13 - 14 | 2470 | 69 | ||
| XM_022415682.1 | XP_022271390.1 | 13 - 14 | 2467 | 69 | ||
| XM_022415683.1 | XP_022271391.1 | 13 - 14 | 2467 | 69 | ||
| XM_022415684.1 | XP_022271392.1 | 14 - 15 | 2581 | 69 | ||
| XM_022415685.1 | XP_022271393.1 | 13 - 14 | 2422 | 69 | ||
| XM_022415686.1 | XP_022271394.1 | 13 - 14 | 2389 | 69 | ||
| XM_022415687.1 | XP_022271395.1 | 13 - 14 | 2386 | 69 | ||
| XM_022415688.1 | XP_022271396.1 | 13 - 14 | 2482 | 69 | ||
| XM_022415689.1 | XP_022271397.1 | 12 - 13 | 2344 | 69 | ||
| XM_022415690.1 | XP_022271398.1 | 12 - 13 | 2341 | 69 | ||
| XM_022415691.1 | XP_022271399.1 | 12 - 13 | 2263 | 69 | ||
| XM_022415692.1 | XP_022271400.1 | 12 - 13 | 2260 | 69 | ||
More Information
Set Membership: |
|

MAP7D3
Genomic Map