Assay Details
| Gene Symbol: NDUFB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005640524.3 | XP_005640581.1 | 2 - 3 | 295 | 65 | |
| XM_022415251.1 | XP_022270959.1 | 3 - 4 | 352 | 65 | ||
More Information
Set Membership: |
|

NDUFB3
Genomic Map