Assay Details
| Gene Symbol: CBY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005625875.2 | XP_005625932.1 | 4 - 5 | 537 | 75 | |
| XM_014117362.2 | XP_013972837.1 | 4 - 5 | 540 | 75 | ||
| XM_022424445.1 | XP_022280153.1 | 3 - 4 | 284 | 75 | ||
| XM_538373.6 | XP_538373.3 | 3 - 4 | 324 | 75 | ||
More Information
Set Membership: |
|

CBY1
Genomic Map