Assay Details
| Gene Symbol: IL11RA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005626742.3 | XP_005626799.1 | 2 - 3 | 252 | 111 | |
| XM_022425538.1 | XP_022281246.1 | 2 - 3 | 253 | 111 | ||
| XM_022425539.1 | XP_022281247.1 | 2 - 3 | 246 | 111 | ||
More Information
Set Membership: |
|

IL11RA
Genomic Map