Assay Details
Gene Symbol: MANEA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005627652.3 | XP_005627709.1 | 4 - 5 | 866 | 96 | |
XM_005627653.3 | XP_005627710.1 | 3 - 4 | 818 | 96 | ||
XM_005627654.3 | XP_005627711.1 | 3 - 4 | 772 | 96 | ||
XM_022426362.1 | XP_022282070.1 | 4 - 5 | 988 | 96 | ||
XM_022426363.1 | XP_022282071.1 | 4 - 5 | 956 | 96 | ||
XM_022426364.1 | XP_022282072.1 | 4 - 5 | 904 | 96 | ||
XM_539046.6 | XP_539046.3 | 3 - 4 | 899 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
