Assay Details
| Gene Symbol: CA14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003639624.4 | XP_003639672.2 | 9 - 10 | 1039 | 65 | |
| XM_022404958.1 | XP_022260666.1 | 9 - 10 | 1054 | 65 | ||
| XM_022404959.1 | XP_022260667.1 | 8 - 9 | 1030 | 65 | ||
| XM_022404960.1 | XP_022260668.1 | 8 - 9 | 874 | 65 | ||
| XM_022404961.1 | XP_022260669.1 | 7 - 8 | 850 | 65 | ||
More Information
Set Membership: |
|

CA14
Genomic Map