Assay Details
| Gene Symbol: MIOX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014117256.1 | XP_013972731.1 | 9 - 10 | 793 | 72 | |
| XM_014117257.1 | XP_013972732.1 | 9 - 10 | 730 | 72 | ||
More Information
Set Membership: |
|

MIOX
Genomic Map