Assay Details
| Gene Symbol: DLA-DMA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005627068.3 | XP_005627125.1 | 2 - 3 | 714 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DLA-DMA| Gene Symbol: DLA-DMA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005627068.3 | XP_005627125.1 | 2 - 3 | 714 | 63 | |
Set Membership: |
|