Assay Details
| Gene Symbol: NFKBIB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005616607.3 | XP_005616664.1 | 3 - 4 | 583 | 61 | |
| XM_541633.5 | XP_541633.1 | 3 - 4 | 790 | 61 | ||
More Information
Set Membership: |
|

NFKBIB
Genomic Map