Assay Details
| Gene Symbol: MORN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005636181.3 | XP_005636238.1 | 435 | 69 | ||
| XM_005636182.3 | XP_005636239.1 | 429 | 69 | |||
| XM_014108082.2 | XP_013963557.1 | 540 | 69 | |||
| XM_014108083.2 | XP_013963558.1 | 541 | 69 | |||
| XM_014108084.2 | XP_013963559.1 | 542 | 69 | |||
| XM_022410218.1 | XP_022265926.1 | 543 | 69 | |||
| XM_022410219.1 | XP_022265927.1 | 542 | 69 | |||
| XM_022410220.1 | XP_022265928.1 | 377 | 69 | |||
More Information
Set Membership: |
|

MORN3
Genomic Map