Assay Details
| Gene Symbol: LOC100855640 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003640150.4 | XP_003640198.1 | 1 - 2 | 735 | 69 | |
| XM_005640125.3 | XP_005640182.1 | 1 - 2 | 734 | 69 | ||
| XM_022414583.1 | XP_022270291.1 | 1 - 2 | 735 | 69 | ||
More Information
Set Membership: |
|

LOC100855640
Genomic Map