Assay Details
| Gene Symbol: LOC486353 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_543479.5 | XP_543479.2 | 2 - 3 | 194 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC486353| Gene Symbol: LOC486353 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_543479.5 | XP_543479.2 | 2 - 3 | 194 | 75 | |
Set Membership: |
|