Assay Details
| Gene Symbol: CCDC113 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014109184.2 | XP_013964659.1 | 2 - 3 | 454 | 93 | |
| XM_022410745.1 | XP_022266453.1 | 2 - 3 | 454 | 93 | ||
| XM_022410753.1 | XP_022266461.1 | 2 - 3 | 454 | 93 | ||
| XM_022410759.1 | XP_022266467.1 | 2 - 3 | 454 | 93 | ||
| XM_022410761.1 | XP_022266469.1 | 2 - 3 | 454 | 93 | ||
| XM_544381.5 | XP_544381.2 | 2 - 3 | 339 | 93 | ||
More Information
Set Membership: |
|

CCDC113
Genomic Map