Assay Details
| Gene Symbol: TRMT10A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005639223.3 | XP_005639280.1 | 2 - 3 | 337 | 88 | |
| XM_005639224.3 | XP_005639281.1 | 3 - 4 | 391 | 88 | ||
| XM_005639227.3 | XP_005639284.1 | 2 - 3 | 1482 | 88 | ||
| XM_022413422.1 | XP_022269130.1 | 2 - 3 | 337 | 88 | ||
| XM_022413423.1 | XP_022269131.1 | 4 - 5 | 463 | 88 | ||
| XM_022413424.1 | XP_022269132.1 | 3 - 4 | 402 | 88 | ||
| XM_544994.6 | XP_544994.2 | 2 - 3 | 792 | 88 | ||
More Information
Set Membership: |
|

TRMT10A
Genomic Map