Assay Details
| Gene Symbol: LACTB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022412246.1 | XP_022267954.1 | 4 - 5 | 496 | 120 | |
| XM_544119.6 | XP_544119.2 | 4 - 5 | 652 | 120 | ||
More Information
Set Membership: |
|

LACTB2
Genomic Map