Assay Details
| Gene Symbol: LOC488215 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014110624.2 | XP_013966099.1 | 2 - 3 | 1629 | 113 | |
| XM_545337.6 | XP_545337.3 | 2 - 3 | 1342 | 113 | ||
More Information
Set Membership: |
|

LOC488215
Genomic Map