Assay Details
| Gene Symbol: PAQR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005638542.3 | XP_005638599.1 | 7 - 8 | 950 | 83 | |
| XM_005638543.3 | XP_005638600.1 | 7 - 8 | 882 | 83 | ||
| XM_005638544.3 | XP_005638601.1 | 6 - 7 | 870 | 83 | ||
| XM_022412540.1 | XP_022268248.1 | 7 - 8 | 983 | 83 | ||
More Information
Set Membership: |
|

PAQR5
Genomic Map