Assay Details
| Gene Symbol: CALHM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003639996.3 | XP_003640044.1 | 1 - 2 | 296 | 129 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003639996.3 transcripts.
Set Membership: |
|

CALHM2
Genomic Map