Assay Details
| Gene Symbol: IFI44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005622026.3 | XP_005622083.1 | 3 - 4 | 700 | 109 | |
| XM_005622027.3 | XP_005622084.1 | 3 - 4 | 700 | 109 | ||
More Information
Set Membership: |
|

IFI44
Genomic Map