Assay Details
| Gene Symbol: TNFRSF13C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_843968.5 | XP_849061.2 | 2 - 3 | 2689 | 148 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TNFRSF13C| Gene Symbol: TNFRSF13C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_843968.5 | XP_849061.2 | 2 - 3 | 2689 | 148 | |
Set Membership: |
|